Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(trifluoromethyl)aniline 98.0+%, TCI America™
SDP

Catalog No. p-7119685 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 328-93-8
Molecular Formula C8H5F6N
Molecular Weight (g/mol) 229.13
MDL Number MFCD00074940
InChI Key XWMVIJUAZAEWIE-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl aniline, 2,5-di trifluoromethyl aniline, 2,5-bis-trifluoromethyl aniline, 2,5-bis-trifluoromethylaniline, benzenamine, 2,5-bis trifluoromethyl, 2,5-bis trifluoromethyl-aniline, 2,5-bis trifluoromethyl benzenamine, 1-amino-2,5-bis trifluoromethyl benzene, 2,5-bis trifluoromethyl phenylamine, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-2,5-xylidine
PubChem CID 719818
IUPAC Name 2,5-bis(trifluoromethyl)aniline
SMILES NC1=CC(=CC=C1C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 177°C
Quantity 5 g
Formula Weight 229.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Bis(trifluoromethyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.