Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorosulfanilic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7126197 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 88-50-6
Molecular Formula C6H5Cl2NO3S
Molecular Weight (g/mol) 242.07
MDL Number MFCD00025159
InChI Key SJCTXIKOXTUQHC-UHFFFAOYSA-N
Synonym 2,5-Dichloroaniline-4-sulfonic Acid
PubChem CID 66619
IUPAC Name 4-amino-2,5-dichlorobenzenesulfonic acid
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)O)Cl)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 242.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorosulfanilic Acid
EINECSNumber (3)-2085
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.