Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-1-(2-thienylmethyl)-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. SP00291DA
Click to view available options
Quantity:

Chemical Identifiers

CAS

306936-14-1

Molecular Formula

C12H13NO2S

Molecular Weight (g/mol)

235.301

MDL Number

MFCD01567217

InChI Key

MYRBXKXOHIDTFU-UHFFFAOYSA-N

Synonym

2,5-dimethyl-1-2-thienylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl pyrrole-3-carboxylic acid, 2,5-dimethyl-1-2-thienylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thien-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-ylmethyl-1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-1-thiophen-2-yl methyl-1h-pyrrole-3-carboxylic acid, maybridge1_008501, 2,5-dimethyl-1-thiophen-2-ylmethyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-1-2-thienylmethyl, 2,5-dimethyl-1-2-thienyl-methyl-1h-pyrrole-3-carboxylic acid

PubChem CID

2736309

IUPAC Name

2,5-dimethyl-1-(thiophen-2-ylmethyl)pyrrole-3-carboxylic acid

SMILES

CC1=CC(=C(N1CC2=CC=CS2)C)C(=O)O

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Quantity

1 g

Formula Weight

235.31

Percent Purity

97%

Chemical Name or Material

2, 5-Dimethyl-1-(2-thienylmethyl)-1H-pyrrole-3-carboxylic acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.