Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-4228387
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24821-22-5
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.11
InChI Key XZNLSDPNMNWCRE-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl benzoic acid, benzoic acid, 2,6-bis trifluoromethyl, 2,6-bis-trifluoromethylbenzoic acid, 2,6-di trifluoromethyl benzoic acid, 2,6-bis-trifluoromethyl-benzoic acid, 2,6-bis trifluoromethyl-benzoic acid, pubchem4988, acmc-1co7v, rarechem al bo 0513, timtec-bb sbb000962
PubChem CID 90618
IUPAC Name 2,6-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C(=C1)C(F)(F)F)C(=O)O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 137.7°C to 138.6°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CO2H
Quantity 1 g
Formula Weight 258.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Bis(trifluoromethyl)benzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 246-479-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.