Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-N-hydroxybenzenecarboximidoyl chloride, Tech., Thermo Scientific™

Catalog No. SPB07103DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 6579-27-7
Packaging Amber Glass Bottle
Molecular Formula C7H4Cl3NO
Quantity 1g
MDL Number MFCD00221426
Synonym 2,6-dichloro-n-hydroxybenzenecarboximidoyl chloride, z-2,6-dichloro-n-hydroxybenzene-1-carbonimidoyl chloride, 2,6-dichloro-n-hydroxybenzimidoyl chloride, 2,6-dichloro-n-hydroxybenzenecarbonimidoyl chloride, hcnhpethchbxchhheefppdql@bx, 2,6-dichloro-alpha-chlorobenzaldoxime, 2,6-dichlorophenyl chloro hydroxyimino methane, 2,6-dichloro-n-hydroxybenzenecarboximidoyl chlorid, e-2,6-dichloro-n-hydroxybenzenecarbonimidoyl chloride, e-2,6-dichloro-n-hydroxybenzene-1-carbonimidoyl chloride
InChI Key LXWQVTNZUIURFE-XFFZJAGNSA-N
SMILES C1=CC(=C(C(=C1)Cl)C(=NO)Cl)Cl
IUPAC Name (1Z)-2,6-dichloro-N-hydroxybenzenecarboximidoyl chloride
Molecular Weight (g/mol) 224.465
PubChem CID 9583163
Formula Weight 224.47
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2, 6-Dichloro-N-hydroxybenzenecarboximidoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.