Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzoic acid, 97%, Thermo Scientific™

Catalog No. SB01105EA
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 385-00-2
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00002411
InChI Key ONOTYLMNTZNAQZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-difluoro, 2,6-difluorobenzoicacid, 2,6-difluoro benzoic acid, 2,6-difluoro-benzoic acid, pubchem2274, benzoic acid,6-difluoro, acmc-209j0h, 2,6,-difluorobenzoic acid, 3-09-00-01330 beilstein handbook reference, ksc223c7l
PubChem CID 9796
IUPAC Name 2,6-difluorobenzoic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula F2C6H3CO2H
Quantity 10 g
Formula Weight 158.1
Percent Purity 97%
Chemical Name or Material 2, 6-Difluorobenzoic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.