Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Pyridinedicarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. p-4228890
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5431-44-7
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
InChI Key PMWXGSWIOOVHEQ-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde #
PubChem CID 79485
IUPAC Name pyridine-2,6-dicarbaldehyde
SMILES C1=CC(=NC(=C1)C=O)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 123.3°C to 124.5°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 135.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 6-Pyridinedicarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 226-589-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.