Learn More
AdipoGen 2 7-Dichlorofluorescein
SDP
Supplier:  AdipoGen CDXD0004G025
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 76-54-0. Formula C20H10Cl2O5. MW 401.2. Synthetic Fluorescent dye. Spectral data Extinction 504nm/Emmission 529nm, in 0.1 M Tris pH 8.0. Used for the determination of carbohydrates by fluorescence densitometry after TLC, in the determination of H2O2 by chemiluminescence, and as a fluorescent additive used in indirect fluorimetric detection in OTC-HPLC. Used as a component/substrate of the 2,7-dichlorofluorescein diacetate DCFH-DA assay to quantitate reactive oxygen species ROS.
Catalog No. 50-256-1329
missing translation for 'QuantityLeft'