Learn More
AdipoGen 2 7-Dichlorofluorescin DA
SDP
Supplier:  AdipoGen CDXD0064M050
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 4091-99-0. Formula C24H16Cl2O7. MW 487.29. Dichlorofluorescin diacetate H2DCFDA is a chemically reduced analog of fluorescein, used as an indicator for reactive oxygen species ROS in cells. The cell permeable non-fluorescent H2DCFDA is cleaved by cellular esterases and oxidation, and converted to the highly fluorescent 2,7-dichlorofluorescein. H2DCFDA might be reactive toward a broad range of oxidizing reactions that may be increased during intracellular oxidant stress. Peroxynitrite is an efficient mediator of this oxidation and neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DCFH. This probe is widely used to monitoring cellular redox processes. Used as a component/substrate of the 2,7-dichlorofluorescein diacetate DCFH-DA assay to quantitate reactive oxygen species ROS. Can detect reactive oxygen species in live cells. Spectral data lambdaEx=504nm/lambdaEm=529nm.
Catalog No. 50-256-1271