Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,9-Dichloro-1,10-phenanthroline 97.0+%, TCI America™
SDP

Catalog No. p-7128145 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 29176-55-4
Molecular Formula C12H6Cl2N2
Molecular Weight (g/mol) 249.09
MDL Number MFCD02084186
InChI Key DNKGIDURJINUOA-UHFFFAOYSA-N
PubChem CID 355196
IUPAC Name 2,9-dichloro-1,10-phenanthroline
SMILES ClC1=CC=C2C=CC3=CC=C(Cl)N=C3C2=N1

Specifications

Melting Point 235°C
Quantity 200 mg
Formula Weight 249.09
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,9-Dichloro-1,10-phenanthroline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.