Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamidobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. p-7118151 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 13493-47-5
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD06739581
InChI Key OWMJAQBUFVTERI-UHFFFAOYSA-N
PubChem CID 326664
IUPAC Name N-(2-formylphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1C=O

Specifications

Melting Point 67°C
Color Yellow
Boiling Point 103°C
Quantity 200 mg
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamidobenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.