Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2-thioxoethyl pivalate, 97%, Thermo Scientific™

Catalog No. RF02972EA
Click to view available options
Quantity:

Chemical Identifiers

CAS

175204-79-2

Molecular Formula

C7H13NO2S

Molecular Weight (g/mol)

175.25

MDL Number

MFCD00204238

InChI Key

COULAOZTCJTHOX-UHFFFAOYSA-N

Synonym

2-amino-2-thioxoethyl pivalate, 2-amino-2-thioxoethyl 2,2-dimethylpropanoate, carbamothioylmethyl 2,2-dimethylpropanoate, 2-tert-butylcarbonyloxy thioacetamide, propanoicacid, 2,2-dimethyl-, 2-amino-2-thioxoethyl ester, propanoic acid, 2,2-dimethyl-, 2-amino-2-thioxoethyl ester, maybridge1_006303, 2-pivaloyloxy-thioacetamide, 2-amino-2-thioxoethylpivalate

PubChem CID

2782113

IUPAC Name

(2-amino-2-sulfanylideneethyl) 2,2-dimethylpropanoate

SMILES

CC(C)(C)C(=O)OCC(N)=S

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Linear Formula

(CH3)3CCO2CH2C(S)NH2

Quantity

10 g

Formula Weight

175.25

Percent Purity

97%

Chemical Name or Material

2-Amino-2-thioxoethyl pivalate

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.