Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2-thioxoethyl pivalate, 97%, Thermo Scientific™

Catalog No. p-3586653
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 175204-79-2
Molecular Formula C7H13NO2S
Molecular Weight (g/mol) 175.25
MDL Number MFCD00204238
InChI Key COULAOZTCJTHOX-UHFFFAOYSA-N
Synonym 2-amino-2-thioxoethyl pivalate, 2-amino-2-thioxoethyl 2,2-dimethylpropanoate, carbamothioylmethyl 2,2-dimethylpropanoate, 2-tert-butylcarbonyloxy thioacetamide, propanoicacid, 2,2-dimethyl-, 2-amino-2-thioxoethyl ester, propanoic acid, 2,2-dimethyl-, 2-amino-2-thioxoethyl ester, maybridge1_006303, 2-pivaloyloxy-thioacetamide, 2-amino-2-thioxoethylpivalate
PubChem CID 2782113
IUPAC Name (2-amino-2-sulfanylideneethyl) 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCC(N)=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula (CH3)3CCO2CH2C(S)NH2
Quantity 1 g
Formula Weight 175.25
Percent Purity 97%
Chemical Name or Material 2-Amino-2-thioxoethyl pivalate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.