Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,5-dibromopyrazine 98.0+%, TCI America™
SDP

Catalog No. p-7117590 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 24241-18-7
Molecular Formula C4H3Br2N3
Molecular Weight (g/mol) 252.90
MDL Number MFCD00673150
InChI Key DTLBKXRFWUERQN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyrazine, 3,5-dibromo-2-pyrazinamine, 3,5-dibromopyrazin-2-ylamine, 3,5-dibromopyrazine-2-ylamine, amino-3,5-dibromopyrazine, pyrazinamine, 3,5-dibromo, 3,5-dibromopyrazine-2-amine, 2-amino-3,5-dibromo pyrazine, 2-amino-3,5-dibromo-pyrazine, 2-pyrazinamine, 3,5-dibromo
PubChem CID 620004
IUPAC Name 3,5-dibromopyrazin-2-amine
SMILES NC1=NC=C(Br)N=C1Br

Specifications

Melting Point 114°C
Quantity 1 g
Formula Weight 252.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,5-dibromopyrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.