Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-(trifluoromethyl)pyrimidine 98.0+%, TCI America™
SDP

Catalog No. p-7117954 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 16075-42-6
Molecular Formula C5H4F3N3
Molecular Weight (g/mol) 163.10
MDL Number MFCD00828624
InChI Key NKOTXYPTXKUCDL-UHFFFAOYSA-N
Synonym 2-amino-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-amine, 2-amino-4-trifluoromethylpyrimidine, 2-pyrimidinamine, 4-trifluoromethyl, 4-trifluoromethyl-2-pyrimidinamine, 4-trifluoromethyl-pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidine-2-ylamine
PubChem CID 595915
IUPAC Name 4-(trifluoromethyl)pyrimidin-2-amine
SMILES NC1=NC=CC(=N1)C(F)(F)F

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 1 g
Formula Weight 163.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyrimidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.