Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromothiazole Hydrobromide 95.0+%, TCI America™
SDP

Catalog No. p-7117523 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 61296-22-8
Molecular Formula C3H4Br2N2S
Molecular Weight (g/mol) 259.947
MDL Number MFCD00012712
InChI Key NUSVDASTCPBUIP-UHFFFAOYSA-N
Synonym 2-amino-5-bromothiazole hydrobromide, 2-amino-5-bromothiazole monohydrobromide, 5-bromothiazol-2-amine hydrobromide, 5-bromo-1,3-thiazol-2-amine hydrobromide, 5-bromo-thiazol-2-ylamine hydrobromide, 2-amino-5-bromothiazole hbr, 2-amino-5-bromo-1,3-thiazole hydrobromide, 2-thiazolamine, 5-bromo-, monohydrobromide, 5-bromo-1,3-thiazole-2-ylamine, bromide, acmc-209mr7
PubChem CID 2723848
IUPAC Name 5-bromo-1,3-thiazol-2-amine;hydrobromide
SMILES C1=C(SC(=N1)N)Br.Br

Specifications

Melting Point 163°C
Quantity 5 g
Formula Weight 259.95
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromothiazole Hydrobromide
RTECSNumber XJ1262200
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.