Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-m-cresol 98.0+%, TCI America™
SDP

Catalog No. p-7117604 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2835-97-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00075082
InChI Key FEDLEBCVFZMHBP-UHFFFAOYSA-N
Synonym 2-amino-m-cresol, phenol, 2-amino-3-methyl, 2-amino-3-methyl-phenol, 2-hydroxy-6-methylaniline, amino-m-cresol, ccris 8319, 2-azanyl-3-methyl-phenol, 2-hydroxy-6-methyl-aniline, acmc-1cp72, dsstox_cid_29117
PubChem CID 3014138
IUPAC Name 2-amino-3-methylphenol
SMILES CC1=CC=CC(O)=C1N

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-m-cresol
EINECSNumber (3)-0707
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.