Learn More
2-Aminoadenosine, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Molecular Formula | C6H5ClFN |
Synonym | benzenamine, 2-chloro-4-fluoro, 2-chloro-4-fluorobenzenamine, 2-chloro-4-fluorophenylamine, 2-chloro-4-fluoro-phenylamine, pubchem2943, 2-chloro-4fluoroaniline, 2-chloro-4-fluroaniline, timtec-bb sbb004110, 4-fluoro-2-chloroaniline, acmc-1cj1h |
InChI Key | XRAKCYJTJGTSMM-UHFFFAOYSA-N |
SMILES | C1=CC(=C(C=C1F)Cl)N |
IUPAC Name | 2-chloro-4-fluoroaniline |
Quantity | 1 g |
Molecular Weight (g/mol) | 145.56 |
PubChem CID | 75013 |
Formula Weight | 282.26 |
Percent Purity | 97% |
Show More |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.