Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzotriazole 98.0+%, TCI America™
SDP

Catalog No. p-7117533 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 1614-11-5
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.142
MDL Number MFCD00724002
InChI Key YYZVLEJDGSWXII-UHFFFAOYSA-N
Synonym 2-Benzotriazolamine
PubChem CID 295514
IUPAC Name benzotriazol-2-amine
SMILES C1=CC2=NN(N=C2C=C1)N

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 100 mg
Formula Weight 134.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzotriazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.