Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenyl Phenyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. p-7117609 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 4273-98-7
Molecular Formula C12H11NO2S
Molecular Weight (g/mol) 233.285
InChI Key JBCUKQQIWSWEOK-UHFFFAOYSA-N
Synonym 2-phenylsulfonyl aniline, 2-amino diphenyl sulfone, 2-aminophenyl phenyl sulfone, 2-benzenesulfonyl aniline, 2-aminodiphenylsulfone, benzenamine, 2-phenylsulfonyl, o-aminodiphenyl sulfone, 2-aminodiphenyl sulfone, unii-6vu39id7lu, 6vu39id7lu
PubChem CID 77956
IUPAC Name 2-(benzenesulfonyl)aniline
SMILES C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2N

Specifications

Melting Point 119°C
Quantity 25 g
Formula Weight 233.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenyl Phenyl Sulfone
EINECSNumber (3)-2164
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.