Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminothiazoline Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. p-7117141 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3882-98-2
Molecular Formula C3H7ClN2S
Molecular Weight (g/mol) 138.613
MDL Number MFCD00012709
InChI Key CODJLHDDIDUKMD-UHFFFAOYSA-N
PubChem CID 520926
IUPAC Name 4,5-dihydro-1,3-thiazol-2-amine;hydrochloride
SMILES C1CSC(=N1)N.Cl

Specifications

Melting Point 200°C
Quantity 5 g
Formula Weight 138.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminothiazoline Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.