Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Anthraceneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7117601 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
250 mg
1 g
5 g

Chemical Identifiers

CAS 141981-64-8
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD07784001
InChI Key PKWBMOXZIMVOJT-UHFFFAOYSA-N
PubChem CID 9855952
IUPAC Name (anthracen-2-yl)boronic acid
SMILES OB(O)C1=CC=C2C=C3C=CC=CC3=CC2=C1

Specifications

Quantity 5 g
Formula Weight 222.05
Physical Form Crystalline Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.