Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-APB, Tocris Bioscience™
GSA_VA

Catalog No. p-4783159 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

IP3 receptor antagonist. Also TRP channel modulator

A functional and membrane permeable D-myo-inositol 1,4,5-trisphosphate (IP3) receptor antagonist (IC50 = 42 μM). Stimulates store-operated calcium (SOC) release at low concentrations (< 10 μM) and inhibits it at higher concentrations (up to 50 μM). Increases STIM-Orai channel conductance and limits ion selectivity. Modulator of TRP channels; blocks TRPC1, TRPC3, TRPC5, TRPC6, TRPV6, TRPM3, TRPM7, TRPM8 and TRPP2 and at higher concentrations stimulates TRPV1, TRPV2 and TRPV3. Also blocks specific gap channel subtypes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO
Molecular Weight (g/mol) 225.10
MDL Number MFCD00014823
InChI Key BLZVCIGGICSWIG-UHFFFAOYSA-N
Synonym 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl
PubChem CID 1598
IUPAC Name 2-aminoethyl diphenylborinate
SMILES NCCOB(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 10 mg
Formula Weight 225.1
Percent Purity >98%
Chemical Name or Material 2-APB
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.