Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzothiazoleacetonitrile, 98%, Thermo Scientific Chemicals

Catalog No. p-7081392
Click to view available options
Quantity:
5 g
25 g

2-Benzothiazoleacetonitrile is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzothiazoleacetonitrile is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dye stuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents .

Chemical Identifiers

CAS 56278-50-3
Molecular Formula C9H6N2S
Molecular Weight (g/mol) 174.221
MDL Number MFCD00051633
InChI Key ZMZSYUSDGRJZNT-UHFFFAOYSA-N
Synonym benzothiazole-2-acetonitrile, 2-benzo d thiazol-2-yl acetonitrile, 2-benzothiazoleacetonitrile, 2-1,3-benzothiazol-2-yl acetonitrile, benzothiazole-2-yl-acetonitrile, 1,3-benzothiazol-2-ylacetonitrile, benzothiazol-2-ylacetonitrile, 2-cyanomethyl-1,3-benzothiazole, 2-benzothiazol-2-ylethanenitrile, 2-cyanomethylbenzothiazole
PubChem CID 342465
IUPAC Name 2-(1,3-benzothiazol-2-yl)acetonitrile
SMILES C1=CC=C2C(=C1)N=C(S2)CC#N

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
UN Number UN3439
Beilstein 128460
Solubility Information Slightly soluble in water.
Formula Weight 174.23
Percent Purity 98%
Chemical Name or Material 2-Benzothiazoleacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.