Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzylacrylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7122930 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5669-19-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00854174
InChI Key RYNDYESLUKWOEE-UHFFFAOYSA-N
Synonym 2-Methylene-3-phenylpropionic Acid
PubChem CID 303571
IUPAC Name 2-benzylprop-2-enoic acid
SMILES OC(=O)C(=C)CC1=CC=CC=C1

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylacrylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.