Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Biphenyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. p-7131448 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 17337-13-2
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.22
MDL Number MFCD00037087
InChI Key IHHUGFJSEJSCGE-UHFFFAOYSA-N
Synonym 2-biphenylyl isocyanate, 2-biphenylisocyanate, 2-biphenyl isocyanate, 2-isocyanatobiphenyl, 2-isocyanato-biphenyl, 2-biphenylylisocyanate, isocyanic acid 2-biphenyl ester, 1,1'-biphenyl, isocyanato, 1-isocyanato-2-phenyl-menzene, 1,1-biphenyl-2-yl isocyanate
PubChem CID 222280
IUPAC Name 2-isocyanato-1,1'-biphenyl
SMILES O=C=NC1=CC=CC=C1C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
UN Number 2206
Formula Weight 195.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Biphenyl Isocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.