Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-chloro-4-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog No. p-7077728
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31928-46-8
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.35
MDL Number MFCD07780667
InChI Key KSCPVCBJIZDZER-UHFFFAOYSA-N
Synonym 1-bromo-2-chloro-5-iodobenzene, 2-bromo-1-chloro-4-iodo-benzene, benzene, 2-bromo-1-chloro-4-iodo, acmc-209hpp, 3-bromo-4-chloroiodobenzene, 3-bromo-4-chloro-1-iodobenzene, benzene,2-bromo-1-chloro-4-iodo, 2-bromanyl-1-chloranyl-4-iodanyl-benzene
PubChem CID 50999503
IUPAC Name 2-bromo-1-chloro-4-iodobenzene
SMILES ClC1=C(Br)C=C(I)C=C1

Specifications

Melting Point 39.0°C to 42.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 317.35
Percent Purity 98%
Chemical Name or Material 2-Bromo-1-chloro-4-iodobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.