Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Bromo-4,4,4-trifluorobutyric acid ethyl ester, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97+% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 249.03 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-4, 4, 4-trifluorobutyric acid ethyl ester |
Safety and Handling
Hazard Category | Warning |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction