Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methylthiophene (stabilized with Copper chip + NaHCO3) 98.0+%, TCI America™
SDP

Catalog No. p-7119106 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 765-58-2
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.059
MDL Number MFCD00130103
InChI Key ACDLOOGOFKSUPO-UHFFFAOYSA-N
Synonym 2-bromo-5-methyl-thiophene, 2-bromo-5-methyl thiophene, thiophene, 2-bromo-5-methyl, 2-methyl-5-bromothiophene, 2-brom-5-methylthiophen, zlchem 358, pubchem5193, acmc-1beki, 2-methyl-5-bromothiphene, 5-bromo-2-methylthiophene
PubChem CID 69831
IUPAC Name 2-bromo-5-methylthiophene
SMILES CC1=CC=C(S1)Br

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
Formula Weight 177.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-methylthiophene (stabilized with Copper chip + NaHCO3)
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.