Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-chloro-4-fluoroaniline 97.0+%, TCI America™
SDP

Catalog No. p-7123100 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 201849-14-1
Molecular Formula C6H4BrClFN
Molecular Weight (g/mol) 224.46
MDL Number MFCD00051707
InChI Key LIBGMUMMWYKJSC-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-6-chloro-4-fluoro, pubchem2922, acmc-209f6h, attercop-chm at130703, 2-bromo-4-fluoro-6-chloroaniline, 2-bromo-6-chloro-4-fluoro-aniline, 2-bromo-6-chloro-4-fluorophenylamine, 6-bromo-2-chloro-4-fluorophenylamine, 2-bromo-6-chloro-4-fluoro-phenylamine, 2-bromo-6-chloro-4-fluoroaniline
PubChem CID 614401
IUPAC Name 2-bromo-6-chloro-4-fluoroaniline
SMILES NC1=C(Cl)C=C(F)C=C1Br

Specifications

Melting Point 57°C
Color Red-White
Quantity 5 g
Formula Weight 224.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-chloro-4-fluoroaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.