Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-7-tert-butylpyrene 98.0+%, TCI America™
SDP

Catalog No. p-7123266 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 78751-80-1
Molecular Formula C20H17Br
Molecular Weight (g/mol) 337.26
InChI Key RYLYZVOQPGVCQT-UHFFFAOYSA-N
PubChem CID 70702341
IUPAC Name 2-bromo-7-tert-butylpyrene
SMILES CC(C)(C)C1=CC2=C3C(=C1)C=CC4=CC(=CC(=C43)C=C2)Br

Specifications

Melting Point 152°C
Color White
Quantity 1 g
Formula Weight 337.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-7-tert-butylpyrene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.