Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-m-xylene 98.0+%, TCI America™
SDP

Catalog No. p-7118605 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 576-22-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000075
InChI Key MYMYVYZLMUEVED-UHFFFAOYSA-N
Synonym 2,6-dimethylbromobenzene, 2-bromo-m-xylene, 1-bromo-2,6-dimethylbenzene, benzene, 2-bromo-1,3-dimethyl, 2,6-dimethyl bromobenzene, m-xylene, 2-bromo, 2,6-dimethylphenyl bromide, 2-bromo-meta-xylene, 2-bromo-1,3-dimethyl-benzene, pubchem3206
PubChem CID 68471
IUPAC Name 2-bromo-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)Br

Specifications

Melting Point -10°C
Boiling Point 206°C
Quantity 25 g
Physical Form Clear Liquid at 20°C
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Bromo-m-xylene
RTECSNumber CY9020000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.