Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoacetylnaphthalene 98.0+%, TCI America™
SDP

Catalog No. p-7118954 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 613-54-7
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
MDL Number MFCD00004109
InChI Key YHXHHGDUANVQHE-UHFFFAOYSA-N
Synonym 2-bromo-2'-acetonaphthone, bromomethyl 2-naphthyl ketone, 2-bromoacetyl naphthalene, 2-bromo-1-naphthalen-2-yl ethanone, 2-bromo-1-2-naphthyl-1-ethanone, ethanone, 2-bromo-1-2-naphthalenyl, 2-bromo-1-2-naphthyl ethanone, 2-bromo-1-naphthalen-2-yl ethan-1-one, alpha-bromo-2'-acetonaphthone, 2-bromo-1-naphthalen-2-yl-ethanone
PubChem CID 69179
IUPAC Name 2-bromo-1-naphthalen-2-ylethanone
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)CBr

Specifications

Melting Point 84°C
Quantity 5 g
UN Number 3261
Formula Weight 249.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoacetylnaphthalene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.