Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butylaniline 98.0+%, TCI America™
SDP

Catalog No. p-7119250 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2696-85-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00130033
InChI Key HDVUPIFFKAHPJY-UHFFFAOYSA-N
PubChem CID 75909
IUPAC Name 2-butylaniline
SMILES CCCCC1=CC=CC=C1N

Specifications

Boiling Point 126°C
Quantity 5 g
Formula Weight 149.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Butylaniline
EINECSNumber (3)-0206
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.