Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1-morpholinoethan-1-one, 95%, Thermo Scientific™

Catalog No. KM10055EA
Click to view available options
Quantity:
1g
10 g

Chemical Identifiers

CAS 1440-61-5
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
MDL Number MFCD00721939
InChI Key YMQRPXBBBOXHNZ-UHFFFAOYSA-N
Synonym 4-2-chloroacetyl morpholine, morpholine, 4-chloroacetyl, 4-chloroacetyl morpholine, n-chloroacetyl morpholine, 2-chloro-1-morpholin-4-yl ethan-1-one, n-chloroacetylmorpholine, chloroacetic acid morpholide, 2-chloro-1-morpholinoethanone, 2-chloro-1-morpholin-4-yl-ethanone, chloroacetic acid, morpholide
PubChem CID 74040
IUPAC Name 2-chloro-1-morpholin-4-ylethanone
SMILES C1COCCN1C(=O)CCl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 163.6
Percent Purity 95%
Chemical Name or Material 2-Chloro-1-morpholinoethan-1-one
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.