Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2-methylbutane 97.0+%, TCI America™
SDP

Catalog No. p-7124275 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 594-36-5
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00000818
InChI Key CRNIHJHMEQZAAS-UHFFFAOYSA-N
Synonym tert-amyl chloride, butane, 2-chloro-2-methyl, 2-methyl-2-chlorobutane, tert-pentyl chloride, 1,1-dimethylpropyl chloride, tertiary pentyl chloride, t-pentyl chloride, acmc-209mdc, 2-chloro-2-methyl butane, 2-chloro-2-methyl-butane
PubChem CID 61143
IUPAC Name 2-chloro-2-methylbutane
SMILES CCC(C)(C)Cl

Specifications

Color Colorless
Boiling Point 86°C
Quantity 25 mL
UN Number 1107
Formula Weight 106.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylbutane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.