Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. SEW01287DA
Click to view available options
Quantity:

Chemical Identifiers

CAS

175205-54-6

Molecular Formula

C7H3Cl2F3O2S

Molecular Weight (g/mol)

279.05

MDL Number

MFCD00052912

InChI Key

NJXDBSSSDPOAFI-UHFFFAOYSA-N

Synonym

2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt

PubChem CID

2736675

SMILES

FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Quantity

1 g

IUPAC Name

2-chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride

Formula Weight

279.06

Percent Purity

97%

Chemical Name or Material

2-Chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Precautionary Statement

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.