Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals

Catalog No. AC453780250
Click to view available options
Quantity:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

2401-24-3

Molecular Formula

C7H8ClNO

Molecular Weight (g/mol)

157.6

InChI Key

GBOUQGUQUUPGLO-UHFFFAOYSA-N

Synonym

6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038

PubChem CID

75460

IUPAC Name

2-chloro-5-methoxyaniline

SMILES

COC1=CC(=C(C=C1)Cl)N

Specifications

Melting Point

23.0°C to 27.0°C

Density

1.2610g/mL

Boiling Point

135.0°C to 137.0°C (12.0 mmHg)

Flash Point

>110°C

Infrared Spectrum

Authentic

Assay Percent Range

97.5% min. (GC)

Refractive Index

1.588

Quantity

25 g

Specific Gravity

1.261

Formula Weight

157.6

Percent Purity

98%

Chemical Name or Material

2-Chloro-5-methoxyaniline

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

EINECSNumber

219-277-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.