Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitrobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7125243 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 16588-02-6
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.563
MDL Number MFCD00007292
InChI Key ZGILLTVEEBNDOB-UHFFFAOYSA-N
Synonym benzonitrile, 2-chloro-5-nitro, 2-chlor-5-nitrobenzonitril german, 5-nitro-2-chlorobenzonitrile, chembl52706, 2-chloro-5-nitro-benzonitrile, 2-chloro-5-nitrobenzenecarbonitrile, 4-chloro-3-cyano-1-nitrobenzene, 2-chlor-5-nitrobenzonitril, pubchem4740, 2-chloro-5-nitrobenzonitril
PubChem CID 85504
IUPAC Name 2-chloro-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)Cl

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 182.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzonitrile
EINECSNumber (3)-1781
RTECSNumber DI3040000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.