Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-fluorophenylboronic acid, 97%, Thermo Scientific™

Catalog No. AC437680050
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 313545-32-3
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD04039892
InChI Key NXSZSZJWZVLHAY-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorophenyl boronic acid, 2-chloro-6-fluorobenzeneboronic acid, 2-chloro-6-fluorophenylboronicacid, boronic acid, 2-chloro-6-fluorophenyl, boronic acid,b-2-chloro-6-fluorophenyl, boronic acid, b-2-chloro-6-fluorophenyl, pubchem18883, acmc-1ad0v, 2-borono-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzene boronic acid
PubChem CID 3549395
IUPAC Name (2-chloro-6-fluorophenyl)boronic acid
SMILES OB(O)C1=C(F)C=CC=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129.9°C to 132.8°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 174.37
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-6-fluorophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.