Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methylpyrazine 98.0+%, TCI America™
SDP

Catalog No. p-7125649 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 38557-71-0
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00055032
InChI Key CKUVSPQGYLELRG-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-pyrazine, pyrazine, 2-chloro-6-methyl, 6-chloro-2-methylpyrazine, 2-chloro-6-methyl-1,4-diazine, 2-methyl-6-chloropyrazine, pubchem18034, acmc-1afqe, 2-chloro-6 methylpyrazine, ksc497m1j
PubChem CID 7170095
IUPAC Name 2-chloro-6-methylpyrazine
SMILES CC1=CN=CC(Cl)=N1

Specifications

Melting Point 52°C
Color White
Boiling Point 83°C
Quantity 1 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylpyrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.