Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methylpyrimidine-4,5-diamine, ≥95%, Thermo Scientific™

Catalog No. RJC03992DE
Click to view available options
Quantity:

Chemical Identifiers

CAS

63211-98-3

Molecular Formula

C5H7ClN4

Molecular Weight (g/mol)

158.59

MDL Number

MFCD01764685

InChI Key

KSGKEVCHVYUUDN-UHFFFAOYSA-N

Synonym

2-chloro-4,5-diamino-6-methylpyrimidine, 4,5-diamino-2-chloro-6-methylpyrimidine, 2-chloro-6-methyl-pyrimidine-4,5-diamine, 2-chloranyl-6-methyl-pyrimidine-4,5-diamine

PubChem CID

275295

IUPAC Name

2-chloro-6-methylpyrimidine-4,5-diamine

SMILES

CC1=NC(Cl)=NC(N)=C1N

Specifications

CAS Min %

95.0

Assay Percent Range

95%

Quantity

5 g

Formula Weight

158.59

Percent Purity

95%

Chemical Name or Material

2-Chloro-6-methylpyrimidine-4, 5-diamine

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.

Precautionary Statement

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.