Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N6-cyclopentyladenosine, Tocris Bioscience™

Catalog No. p-4783684 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective A1 agonist

Potent and selective adenosine A1 receptor agonist (Ki values are 0.8, 2300 and 42 nM for human A1, A2A and A3 receptors respectively; EC50 = 18800 nM for hA2B). Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 37739-05-2
Molecular Formula C15H20ClN5O4
Molecular Weight (g/mol) 369.81
MDL Number MFCD00078574
InChI Key XSMYYYQVWPZWIZ-IDTAVKCVSA-N
Synonym ccpa, 2-chloro-n6-cyclopentyladenosine, 2-chloro-n-cyclopentyladenosine, 2-chloro-n 6 cyclopentyladenosine, ccpa biochemistry, adenosine, 2-chloro-n-cyclopentyl, 3h ccpa, 3h-ccpa, 2r,3r,4s,5r-2-2-chloro-6-cyclopentylamino purin-9-yl-5-hydroxymethyl oxolane-3,4-diol, 2-chloro-n6-cyclopentyladenosine hemihydrate
PubChem CID 123807
IUPAC Name (2R,3R,4S,5R)-2-[2-chloro-6-(cyclopentylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC3CCCC3)N=C(Cl)N=C12

Specifications

Quantity 10 mg
Formula Weight 369.81
Percent Purity >98%
Chemical Name or Material 2-Chloro-N6-cyclopentyladenosine
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.