Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Chlorophenyl)methanesulfonyl chloride, ≥97%, Thermo Scientific™

Catalog No. MO07829DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 77421-13-7
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.08
MDL Number MFCD04116369
InChI Key CHPZYFXSICSCNY-UHFFFAOYSA-N
Synonym 2-chlorophenyl methanesulfonyl chloride, 2-chloro-phenyl-methanesulfonyl chloride, chloro 2-chlorophenyl methyl sulfone, pubchem5500, acmc-1bfc0, 2-chlorobenzylsulfonyl chloride, benzenemethanesulfonylchloride, 2-chloro, 2-chloro-phenyl methanesulfonyl chloride, 2-chlorophenyl methane-sulfonyl chloride
PubChem CID 2757801
IUPAC Name (2-chlorophenyl)methanesulfonyl chloride
SMILES ClC1=CC=CC=C1CS(Cl)(=O)=O

Specifications

CAS Min % 97.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material (2-Chlorophenyl)methanesulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.