Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cl-IB-MECA, Tocris Bioscience™

Catalog No. p-4783162 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective A3 agonist

High affinity and extremely selective A3 adenosine receptor agonist (Ki = 0.33 nM). Displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively. Exhibits high selectivity over the Na+-independent adenosine transporter.

Chemical Identifiers

CAS 163042-96-4
Molecular Formula C18H18ClIN6O4
Molecular Weight (g/mol) 544.734
InChI Key IPSYPUKKXMNCNQ-PFHKOEEOSA-N
Synonym 2-cl-ib-meca, cl-ib-meca, c-ibza-mu, namodenoson, ci-ib-meca, 2s,3s,4r,5r-5-2-chloro-6-3-iodobenzyl amino-9h-purin-9-yl-3,4-dihydroxy-n-methyltetrahydrofuran-2-carboxamide, unii-z07jr07j6c, 2cl-ib-meca, 2-chloro-n 6-3-iodobenzyl adenosine-5'-n-methyluronamide, 1-2-chloro-6-3-iodophenyl methyl amino-9h-purin-9-yl-1-deoxy-n-methyl-beta-d-ribofuranuronamide
PubChem CID 3035850
IUPAC Name (2S,3S,4R,5R)-5-[2-chloro-6-[(3-iodophenyl)methylamino]purin-9-yl]-3,4-dihydroxy-N-methyloxolane-2-carboxamide
SMILES CNC(=O)C1C(C(C(O1)N2C=NC3=C2N=C(N=C3NCC4=CC(=CC=C4)I)Cl)O)O

Specifications

Quantity 10 mg
Formula Weight 544.74
Percent Purity >98%
Chemical Name or Material 2-Cl-IB-MECA
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.