Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-6-hydroxybenzothiazole, 97%, Thermo Scientific Chemicals

Catalog No. AC450940010
Click to view available options
Quantity:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

939-69-5

Molecular Formula

C8H4N2OS

Molecular Weight (g/mol)

176.19

MDL Number

MFCD00296905

InChI Key

SQAVNBZDECKYOT-UHFFFAOYSA-N

Synonym

2-cyano-6-hydroxybenzothiazole, 6-hydroxybenzo d thiazole-2-carbonitrile, 6-hydroxybenzothiazole-2-carbonitrile, 2-benzothiazolecarbonitrile, 6-hydroxy, 6-hydroxy-2-benzothiazolecarbonitrile, pubchem19680, 6-hydroxy-2-cyanobenzothiazole, 6-hydroxy-2-cyano-benzothiazole, 6-hydroxy-benzothiazole-2-carbonitrile, 6-hydroxy-benzo d thiazole-2-carbonitrile

PubChem CID

9881912

IUPAC Name

6-hydroxy-1,3-benzothiazole-2-carbonitrile

SMILES

OC1=CC=C2N=C(SC2=C1)C#N

Specifications

Melting Point

212.0°C to 215.0°C

Infrared Spectrum

Authentic

Assay Percent Range

96% min. (HPLC)

Packaging

Glass Bottle

Quantity

1 g

Formula Weight

176.2

Percent Purity

97%

Chemical Name or Material

2-Cyano-6-hydroxybenzothiazole

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if inhaled.

Precautionary Statement

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.