Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-6-hydroxybenzothiazole, 97%, Thermo Scientific Chemicals

Catalog No. p-7077357
Click to view available options
Quantity:
1 g
5 g

CAS: 939-69-5 | C8H4N2OS | 176.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 939-69-5
Molecular Formula C8H4N2OS
Molecular Weight (g/mol) 176.19
MDL Number MFCD00296905
InChI Key SQAVNBZDECKYOT-UHFFFAOYSA-N
Synonym 2-cyano-6-hydroxybenzothiazole, 6-hydroxybenzo d thiazole-2-carbonitrile, 6-hydroxybenzothiazole-2-carbonitrile, 2-benzothiazolecarbonitrile, 6-hydroxy, 6-hydroxy-2-benzothiazolecarbonitrile, pubchem19680, 6-hydroxy-2-cyanobenzothiazole, 6-hydroxy-2-cyano-benzothiazole, 6-hydroxy-benzothiazole-2-carbonitrile, 6-hydroxy-benzo d thiazole-2-carbonitrile
PubChem CID 9881912
IUPAC Name 6-hydroxy-1,3-benzothiazole-2-carbonitrile
SMILES OC1=CC=C2N=C(SC2=C1)C#N

Specifications

Melting Point 212.0°C to 215.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 176.2
Percent Purity 97%
Chemical Name or Material 2-Cyano-6-hydroxybenzothiazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.