Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2′-Deoxyuridine 99.0+%, TCI America™
SDP

Catalog No. p-7126006 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 951-78-0
Molecular Formula C9H12N2O5
Molecular Weight (g/mol) 228.20
MDL Number MFCD00006527
InChI Key MXHRCPNRJAMMIM-SHYZEUOFSA-N
Synonym 2'-deoxyuridine, deoxyuridine, uracil deoxyriboside, 2-deoxyuridine, 1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, deoxyribose uracil, uridine, 2'-deoxy, 2'-desoxyuridine, unii-w78i7ay22c, ccris 2832
PubChem CID 13712
ChEBI CHEBI:16450
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Melting Point 164°C
Color White
Quantity 1 g
Formula Weight 228.20
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxyuridine
EINECSNumber (9)-2228
RTECSNumber YU7490000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.