Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylphenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. p-7117507 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3056-59-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD02093429
InChI Key VERVERDPMMBDSS-UHFFFAOYSA-N
Synonym Acetic Acid 2-Ethylphenyl Ester
PubChem CID 9898919
IUPAC Name (2-ethylphenyl) acetate
SMILES CCC1=CC=CC=C1OC(=O)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylphenyl Acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.