Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Thermo Scientific™

Catalog No. SEW05200DA
Click to view available options
Quantity:

Chemical Identifiers

CAS

365-12-8

Molecular Formula

C13H8FO2

Molecular Weight (g/mol)

215.20

MDL Number

MFCD00278732

InChI Key

SLKZDWAZOKIEEU-UHFFFAOYSA-M

Synonym

2'-fluorobiphenyl-4-carboxylic acid, 4-2-fluorophenyl benzoic acid, 2'-fluoro-1,1'-biphenyl-4-carboxylic acid, 2'-fluoro 1,1'-biphenyl-4-carboxylic acid, 2'-fluoro-biphenyl-4-carboxylic acid, maybridge3_006504, amtda094, acmc-1cr23, 4-2-fluorophenyl-benzoic acid, 2'-fluoro-4-biphenylcarboxylic acid

PubChem CID

715022

SMILES

[O-]C(=O)C1=CC=C(C=C1)C1=CC=CC=C1F

Specifications

Assay Percent Range

95%

Packaging

Amber Glass Bottle

Linear Formula

FC6H4C6H4CO2H

Quantity

1 g

IUPAC Name

4-(2-fluorophenyl)benzoic acid

Formula Weight

216.21

Percent Purity

95%

Chemical Name or Material

2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.